VWR® AVS TITRINORM Buffer Solution, pH 9 (at 20 °C), Borate free
Lieferant: VWR Chemicals
32037.297EA
46.8
EUR
32037.297
VWR® AVS TITRINORM Buffer Solution, pH 9 (at 20 °C), Borate free
Puffer
pH-Puffer
Solution prepared from AnalaR® NORMAPUR® grade analytical reagents.
- Borate free
- Reference temperature 20 °C
- True analytical value on the label to an accuracy of ±0,02 pH units
- Traceable to SRM from NIST
- Batch number and packing date for traceability
- Certificate of Analysis available at website
Bestellinformation: If significantly large volumes are required please contact your local Avantor sales office for a customised offer.