Natriumhydroxid concentrate for dilution to 1L, 0.25 M NaOH (0.25N) Volumetric, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Caustic soda
32040-1EA
32040-1EAEA
42.08
EUR
32040-1EA
Natriumhydroxid concentrate for dilution to 1L, 0.25 M NaOH (0.25N) Volumetric, Fluka™
Sodium hydroxide
Concentrate for 1L standard solution
Formel:
NaOH Molecular Weight: 40 g/mol Siedepunkt: 105…140 °C (1.013 hPa) Schmelzpunkt: –12 – –10 °C. Dichte: 1.08 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003548 CAS-Nummer: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8 II Merck Index: 14,08627 |
Spezifikation Testergebnisse
Factor | 0.998 - 1.002 |
BAM/EMPA reference material | effective |
BAM/EMPA Certificate Number | effective |
Traceable to NIST SRM | effective |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...