Zinksulfat concentrate for dilution to 1L, 0.1 M ZnSO4 Volumetric, Fluka™

Lieferant: Honeywell Chemicals
Gefahr

32047-1EA
32047-1EAEA 69.66 EUR
32047-1EA
Zinksulfat concentrate for dilution to 1L, 0.1 M ZnSO4 Volumetric, Fluka™
Zinksulfatlösungen

Concentrate for 1L standard solution

Formel: ZnSO₄·7H₂O
Molecular Weight: 287.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149894
CAS-Nummer: 7446-20-0
EINECS: 231-793-3

Bestellen

Spezifikation Testergebnisse

Factor 0.998 - 1.002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR