2,4-Dichlorobenzenediazonium-1,5-naphthalenedisulphonate hydrate, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

320609-5G
320609-5GEA 507 EUR
320609-5G
2,4-Dichlorobenzenediazonium-1,5-naphthalenedisulphonate hydrate, Sigma-Aldrich®
2,4-Dichlorobenzenediazonium-1,5-naphthalenedisulphonate hydrate

Used in spectrophotometric detection of total bilirubin in serum.

Formel: C₁₆H₁₀Cl₂N₂O₆S₂·xH₂O
Molecular Weight: 461.29 g/mol (anhy)
Schmelzpunkt: 195 °C (Zers., Lit.)
Lagertemperatur: Kühlschrank
MDL: MFCD08457580
CAS-Nummer: 123333-91-5
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e24c24c6a7cb1c3fab87a2a5d1a214c1a829957d&vid=bdf21aa19233db0e6c1d4a53ee07d2547092ed4a&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR