VWR® ConvoL NORMADOSE® Buffer Concentrate for Buffer Solution acc. to Weise for 1000 ml, pH 7.20
Lieferant: VWR Chemicals
32075.600EA
35.1
EUR
32075.600
VWR® ConvoL NORMADOSE® Buffer Concentrate for Buffer Solution acc. to Weise for 1000 ml, pH 7.20
Puffer
pH-Puffer
This product is a ConvoL NORMADOSE® concentrate and contains exactly the amount of substance to prepare one liter of solution.