Pesticide reference standard, Posaconazole, VETRANAL(TM), analytical standard, Supelco®
Lieferant: Merck
32103-25MG
32103-25MGEA 106 EUR
32103-25MG
Pesticide reference standard, Posaconazole, VETRANAL(TM), analytical standard, Supelco®
Standards Organische Standards
Posaconazole may have been used as reference standard in HPLC analysis for quantification of posaconazole from human plasma.
  • It is an azole antifungal drug used mostly against zygomycetes
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfa50d69774616341d7d88a6e64feb3b4dbf9e12&vid=dfa50d69774616341d7d88a6e64feb3b4dbf9e12&prid=web&clid=SAPDAM