Gallium(III)bromid, wasserfrei

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Gallium (III) bromide

32107.09
32107.09EA 373 EUR
32107.09
Gallium(III)bromid, wasserfrei
Gallium(III)bromid

CAS No.: 13450-88-9 Molecular Formula: GaBr3 Formula Weight: 309.45 Physical Form: Crystalline Density: 3.69 Sensitivity: Moisture Sensitive Appearance: White Odour: Odourless Melting Point: 122° Boiling Point: 279° UN No.: 3260 MDL No.: MFCD00016100

Formel: Br₃Ga
Molecular Weight: 309,44 g/mol
Siedepunkt: 279 °C
Schmelzpunkt: 122 °C
Dichte: 3,69
Lagertemperatur: Raumtemperatur
MDL: MFCD00016100
CAS-Nummer: 13450-88-9
EINECS: 236-609-5
UN: 3260
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR