Gallium(III)fluorid, wasserfrei ≥99,85% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

32112.06
32112.06EA 465 EUR
32112.06
Gallium(III)fluorid, wasserfrei ≥99,85% (Metall-Basis)
Gallium trifluoride

Purity: 99.85% CAS No.: 7783-51-9 Molecular Formula: GaF3 Formula Weight: 126.72 Physical Form: Powder Density: 4.47 Appearance: White Odour: Odourless Melting Point: 800° subl. Boiling Point: ca 1000° Merck Reference: 14,4354 UN No.: 3260 MDL No.: MFCD00011021

Formel: F₃Ga
Molecular Weight: 126,72 g/mol
Siedepunkt: ∼1000 °C
Schmelzpunkt: 800 °C (Subl.)
Dichte: 4,47
Lagertemperatur: Raumtemperatur
MDL: MFCD00011021
CAS-Nummer: 7783-51-9
EINECS: 232-004-5
UN: 3260
ADR: 8,III
Merck Index: 14,4354

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR