Aluminiumkaliumsulfat Dodecahydrat 98,0-102,0%, körnig, AR® ACS, Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
Synonyme:
Alumen, Kalialaun, Aluminiumkaliumbis(sulfat) Dodecahydrat
3216-20EA
0
EUR
3216-20
Aluminiumkaliumsulfat Dodecahydrat 98,0-102,0%, körnig, AR® ACS, Macron Fine Chemicals™
Aluminiumkaliumsulfat Dodecahydrat
Formel:
AlK(SO₄)₂·12H₂O Molecular Weight: 474,39 g/mol Schmelzpunkt: 92,5 °C Dichte: 1,72 g/cm³ (20 °C) |
MDL:
MFCD00149143 CAS-Nummer: 7784-24-9 EINECS: 233-141-3 Merck Index: 12,00373 |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | 98.0 - 102.0 % |
ACS - Assay (AlK(SO₄)₂·12H₂O) | 98.0 - 102.0 % |
ACS - Ammonium (NH₄) | ≤0.005 % |
ACS - Chloride (Cl) | ≤5 ppm |
ACS - Heavy Metals (as Pb) | ≤0.0005 % |
ACS - Insoluble Matter | ≤0.005 % |
ACS - Iron (Fe) | ≤0.001 % |
ACS - pH of 5% Solution at 25°C | 2.0 - 4.0 |
ACS - Sodium (Na) | ≤0.02 % |
Solubility | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...