Diethylether ≥99,8% enthält Inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr


Stabilisierung: Stabilisiert mit BHT/Ionol/2,6-Di-tert-butyl-4-methylphenol

32203-1L 32203-2.5L 32203-4X2.5L 32203-4X5L 32203-5L 32203-6X1L 32203-27L 32203-500ML 32203-7L-RC
32203-1LEA 94.88 EUR
32203-1L 32203-2.5L 32203-4X2.5L 32203-4X5L 32203-5L 32203-6X1L 32203-27L 32203-500ML HONC32203-7L-RC
Diethylether ≥99,8% enthält Inhibitor, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Diethylether

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 1696894
RTECS Number: KI5775000
UNSPSC Code: 12191602

Formel: (CH₃CH₂)₂O
Siedepunkt: 34…35 °C (1,013 hPa)
Dichte: 0,71 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011646
CAS-Nummer: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (color) complying
Appearance (form) complying
Assay (GC) Min. 99.8 %
Boiling range 34 - 35 °C
Density (D 20/4) 0.713 - 0.714
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.03 %
Free acid (as CH₃COOH) Max. 0.0002 %
Titrable Acid Max. 0.0002 meq/g
Aluminium (Al) Max. 0.50 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.10 ppm
Calcium (Ca) Max. 0.50 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.05 ppm
Magnesium (Mg) Max. 0.10 ppm
Manganese (Mn) Max. 0.02 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.05 ppm
Tin (Sn) Max. 0.10 ppm
Zinc (Zn) Max. 0.05 ppm
Acetone (GC) Max. 0.005 %
Aldehydes complying
Carbonyl compounds (as CO) Max. 0.001 %
Carbonyl Compounds (as HCHO) Max. 0.001 %
Ethanol (GC) Max. 0.02 %
Methanol (GC) Max. 0.02 %
Peroxides (as H₂O₂) Max. 0.3 ppm
Reaction against H₂SO₄ complying
APHA Max. 10
Assay BHT (GC) 5 - 8 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR