Ethanol absolut (versteuert), klar, farblos ≥99,8% (durch GC) Reag. Ph. Eur., ISO, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Ethyl alcohol

32205-160KG 32205-1L 32205-2.5L 32205-2.5L-GL 32205-25L 32205-4X2.5L 32205-6X1L 32205-4X2.5L-G 32205-190L-RC 32205-5L
32205-160KGEA 13695.14 EUR
32205-160KG 32205-1L 32205-2.5L 32205-2.5L-GL 32205-25L 32205-4X2.5L 32205-6X1L 32205-4X2.5L-G 32205-190L-RC HONC32205-5L
Ethanol absolut (versteuert), klar, farblos ≥99,8% (durch GC) Reag. Ph. Eur., ISO, Riedel-de Haën™
Ethanol
Formel: C₂H₅OH
Molecular Weight: 46.07 g/mol
Siedepunkt: 78…79 °C (1.013 hPa)
Schmelzpunkt: –115 °C
Dichte: 0.790…0.791 g/cm3 (20 °C)
Flammpunkt: 12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003568
CAS-Nummer: 64-17-5
UN: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Bestellen

Spezifikation Testergebnisse

Appearance complying
Assay (GC) Min. 99.8 Vol.%
Boiling range 78 - 79 °C
Density (D 20/20) 0.790 - 0.791
Refractive index (n 20/D) 1.3612 - 1.3618
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.2 %
Free acid (as CH₃COOH) Max. 0.0005 %
Free alkali (as NH₃) Max. 0.0001 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Heavy metals (as Pb) Max. 0.0001 %
Acetone (GC) Max. 0.001 %
Aldehydes (as CH₃CHO) Max. 0.001 %
Isoamyl alcohol (GC) Max. 0.05 %
Carbonyl compounds (as CO) Max. 0.003 %
Furfural complying
Methanol (GC) Max. 200 Vol.ppm
2-propanol (GC) Max. 0.003 %
Higher alcohols (GC) Max. 0.01 %
Volatile impurities (GC) complying
KMnO₄ reducing matter (as O) Max. 0.0003 %
Reaction against H₂SO₄ complying
UV-absorption complying
APHA Max. 10
Form liquid (clear, colorless)
Explosion Limit 19 %, 60 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR