5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB), Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem
Achtung

322123-5
322123-5EA 128 EUR
322123-5
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB), Sigma-Aldrich®
5,5'-Dithiobis(2-nitrobenzoesäure) (Ellman's Reagenz, DTNB)
Formel: C₁₄H₈N₂O₈S₂
Schmelzpunkt: 240…244 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007140
CAS-Nummer: 69-78-3
EINECS: 200-714-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR