Methanol ≥99,8% (durch GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Hydroxymethan, Methylalkohol, Holzalkohol

32213-45L 32213-7L 32213-150KG 32213-1L 32213-2.5L 32213-25L 32213-4X2.5L 32213-4X5L 32213-5L 32213-6X1L 32213-45KG 32213-18L 32213-395LRC 32213-395L-RC
32213-45LEA 1764.46 EUR
32213-45L 32213-7L 32213-150KG 32213-1L 32213-2.5L 32213-25L 32213-4X2.5L 32213-4X5L 32213-5L 32213-6X1L 32213-45KG HONC32213-18L HONC32213-395LRC HONC32213-395L-RC
Methanol ≥99,8% (durch GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
Methanol

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 1098229

Formel: CH₃OH
Molecular Weight: 32.04 g/mol
Siedepunkt: 64…65 °C (1.013 hPa)
Schmelzpunkt: –98 °C
Dichte: 0.79 g/cm³ (20 °C)
Flammpunkt: 11 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004595
CAS-Nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (form) complying
Appearance (color) complying
Appearance complying
Assay (GC) Min. 99.8 %
Boiling range 64 - 65 °C
Density (D 20/20) 0.791 - 0.793
Refractive index (n 20/D) 1.328 - 1.330
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as HCOOH) Max. 0.002 %
Free alkali (as NH₃) Max. 0.00005 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000001 %
Iron (Fe) Max. 0.00001 %
Potassium (K) Max. 0.00005 %
Lithium (Li) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000001 %
Molybdenum (Mo) Max. 0.00001 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000002 %
Tin (Sn) Max. 0.00001 %
Strontium (Sr) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Chloride (Cl) Max. 0.00005 %
Sulfate (SO₄) Max. 0.0001 %
KMnO₄ reducing matter (as O) Max. 0.00025 %
Acetone (GC) Max. 0.001 %
Ketones (as CH₃COCH₃) Max. 0.001 %
Aldehydes (as CH₃CHO) Max. 0.001 %
Acetaldehyde Max. 0.001 %
Ethanol (GC) Max. 0.1 %
Formaldehyde Max. 0.001 %
Carbonyl compounds (as CO) Max. 0.005 %
Carbonyl compounds (ACS) complying
Mixable with H₂O complying
Reaction against H₂SO₄ complying
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR