Ammoniaklösung strong NF, ACS, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Ammoniumhydroxid, Ammoniak
3226-20EA
0
EUR
3226-20
3226-45
Ammoniaklösung strong NF, ACS, J.T.Baker®
Ammoniaklösung
Ammonia Solution, Strong; Ammonium Hydroxide
Formel:
NH₃ |
MDL:
MFCD00066650 CAS-Nummer: 1336-21-6 UN: 2672 ADR: 8,III |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | Passes Test |
GMP Manufactured Product | ≤0.02% |
Meets N.F. Requirements | Passes Test |
CAUTION: For Manufacturing, processing or repackaging | 27.0 - 30.0% |
Bulk Pharmaceutical Chemical | 28.0 - 30.0% |
NF - Identification | Passes Test |
NF - Nonvolatile Residue | ≤0.02 % |
NF - Readily Oxidizable Substances | Passes Test |
NF - Assay (NH₃)(NF) | 27.0 - 30.0 % |
ACS - Assay (as NH₃) | 28.0 - 30.0 % |
ACS - Carbon Dioxide (CO₂) | ≤0.002 % |
ACS - Residue after Ignition | ≤0.002 % |
ACS - Substances Reducing Permanganate | Passes Test |
Appearance | Passes Test |
Arsenic and Antimony (as As) | ≤3 ppm |
Chloride (Cl) | ≤0.5 ppm |
Trace Impurities - Heavy Metals | ≤0.5 ppm |
Nitrate (NO₃) | ≤2 ppm |
Phosphate (PO₄) | ≤2 ppm |
Sulfate (SO₄) | ≤2 ppm |
Trace Impurities - Aluminum (Al) | ≤0.300 ppm |
Trace Impurities - Boron (B) | ≤0.100 ppm |
Trace Impurities - Calcium (Ca) | ≤0.300 ppm |
Trace Impurities - Chromium (Cr) | ≤0.200 ppm |
Trace Impurities - Copper (Cu) | ≤0.10 ppm |
Trace Impurities - Gold (Au) | ≤0.300 ppm |
Trace Impurities - Iron (Fe) | ≤0.2 ppm |
Trace Impurities - Lead (Pb) | ≤0.200 ppm |
Trace Impurities - Magnesium (Mg) | ≤0.300 ppm |
Trace Impurities - Manganese (Mn) | ≤0.200 ppm |
Trace Impurities - Nickel (Ni) | ≤0.1 ppm |
Trace Impurities - Potassium (K) | ≤0.300 ppm |
Trace Impurities - Sodium (Na) | ≤2.000 ppm |
Trace Impurities - Tin (Sn) | ≤0.300 ppm |
Trace Impurities - Titanium (Ti) | ≤0.300 ppm |
Trace Impurities - Zinc (Zn) | ≤0.300 ppm |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP (232), EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...