Kaliumpermanganat ≥99,0%, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
3228-07EA
0
EUR
3228-07
Kaliumpermanganat ≥99,0%, BAKER ANALYZED® ACS, J.T.Baker®
Potassium permanganate
Formel:
KMnO₄ |
MDL:
MFCD00011364 CAS-Nummer: 7722-64-7 UN: 1490 ADR: 5.1,II |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | — |
For Laboratory,Research,or Manufacturing Use |
|
Assay (KMnO₄) | ≥99.0 % |
Insoluble Matter | ≤0.2 % |
Chloride and Chlorate (as Cl) | ≤0.005 % |
Sulfate (SO₄) | ≤0.02 % |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...