n-Hexan ≥99% (durch GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Lieferant: Honeywell Chemicals
|
Gefahr
|
32293-1L
32293-2.5L
32293-25L
32293-4X2.5L
32293-6X1L
32293-7L-RC
32293-18L
32293-1LEA
133.08
EUR
32293-1L
32293-2.5L
32293-25L
32293-4X2.5L
32293-6X1L
HONC32293-7L-RC
HONC32293-18L
n-Hexan ≥99% (durch GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
n-Hexan
ACS GENERAL USE SOLVENTS
- Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- Suitable for less demanding applications such as routine isocratic HPLC
- Variations and decrease issues traceable to solvent contamination
- All solvents are tested to meet ACS general use specifications
Beilstein Number: 1730733
RTECS Number: MN9275000
UNSPSC Code: 12191502
Formel:
H₃C(CH₂)₄CH₃ Siedepunkt: 66…69 °C(1,013 hPa) Schmelzpunkt: –95 °C Dichte: 0,660…0.668 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009520 CAS-Nummer: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 13,04712 |
|
Spezifikation Testergebnisse
Refractive Index | n20/D 1.375 (lit.) |
Assay (GC) | Min. 99 % |
Boiling range | 68 - 69 °C |
Boiling range 67-69 °C | Min. 95 % |
Density (D 20/4) | 0.658 - 0.662 |
Density (D 20/20) | 0.659 - 0.663 |
Refractive index (n 20/D) | 1.3745 - 1.3755 |
Non-volatile matter | Max. 0.001 % |
Water (Karl Fischer) | Max. 0.01 % |
Free acid (as CH₃COOH) | Max. 0.002 % |
Aluminium (Al) | Max. 0.00005 % |
Boron (B) | Max. 0.000002 % |
Barium (Ba) | Max. 0.00001 % |
Calcium (Ca) | Max. 0.00005 % |
Cadmium (Cd) | Max. 0.000005 % |
Cobalt (Co) | Max. 0.000002 % |
Chromium (Cr) | Max. 0.000002 % |
Copper (Cu) | Max. 0.000002 % |
Iron (Fe) | Max. 0.00001 % |
Magnesium (Mg) | Max. 0.00001 % |
Manganese (Mn) | Max. 0.000002 % |
Nickel (Ni) | Max. 0.000002 % |
Lead (Pb) | Max. 0.00001 % |
Tin (Sn) | Max. 0.00001 % |
Zinc (Zn) | Max. 0.00001 % |
Aromatic substances (as C₆H₆) | Max. 0.01 % |
S-compounds (as S) | Max. 0.005 % |
Thiophene | Max. 0.0001 % |
Reaction against H₂SO₄ | complying |
APHA | Max. 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...