N,N-Bis(2-hydroxyethyl)ethylenediamine
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
32312.03
32312.03EA
146
EUR
32312.03
N,N-Bis(2-hydroxyethyl)ethylenediamine
N,N-Bis(2-hydroxyethyl)ethylenediamine
CAS No.: 3197-06-6
Molecular Formula: (HOCH2CH2)2NCH2CH2NH2
Formula Weight: 148.21
Physical Form: Liquid
Density: 1.069
Odour: Amine-like
Boiling Point: 174-178°/3-4mm
UN No.: UN2735
MDL No.: MFCD00014822
Formel:
C₆H₁₆N₂O₂ Siedepunkt: 174…178 °C (3…4 mmHg) Dichte: 1,069 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014822 CAS-Nummer: 3197-06-6 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...