4-(2-Pyridylazo)resorcin 96%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 4-(2'-Pyridylazo)resorcinol

M-Clarity™ Qualitätsstufe = MQ200

323209-25G 323209-5G
323209-25GEA 854 EUR
323209-25G 323209-5G
4-(2-Pyridylazo)resorcin 96%, Sigma-Aldrich®
4-(2-Pyridylazo)resorcin

4-(2-Pyridylazo)resorcinol (PAR) is a monosodium salt(1), dibasic acid that forms the protonated complexes with most metal ions.


4-(2-Pyridylazo)resorcinol (PAR) serves as a metallochromic indicator and is suitable as a chromogenic agent for the quantitative determination of over 50 elements.

Formel: C₁₁H₉N₃O₂
Schmelzpunkt: 192,..202 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00044237
CAS-Nummer: 1141-59-9
EINECS: 214-528-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9f6536f522a531b3cdb3fc1413cd80c7371a1a5&vid=557eb6dc79d4239c26bd7e5a55133d1917cd88c1&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (colour) Conforms to requirements
Appearance (form) Powder
Purity (HPLC area %) ≥95.5 %
Carbon content 58.6 - 64.2 %
Nitrogen content 18.6 - 20.4 %
Infrared spectrum Conforms to structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR