Methylgrün-Zinkchlorid-Doppelsalz <0,5% crystal violet 80% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr


M-Clarity™ Qualitätsstufe = MQ200

323829-25G 323829-5G
323829-25GEA 752 EUR
323829-25G 323829-5G
Methylgrün-Zinkchlorid-Doppelsalz <0,5% crystal violet 80% (Farbstoffgehalt), Sigma-Aldrich®
Methylgrün-Zinkchlorid-Doppelsalz

Methyl Green is a triphenylmethane dye which is mainly used in combination with pyronin. It is suitable for the detection of DNA and RNA. Used in the Methyl Green-Pyronin methods to demonstrate DNA and RNA in tissue sections.

Formel: C₂₇H₃₅BrClN₃·xZnCl₂
Schmelzpunkt: >300 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00151094
CAS-Nummer: 7114-03-6
EINECS: 230-415-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa6d9bc0d4c9b4b3bcc376078b8a7e645e7f775f&vid=2ff14c1e48ed1a5c2f6d654486fa30707360555c&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form); Powder or Crystalline Powder Conforms to Requirements
Infrared spectrum Conforms to Structure
Wavelength, c = 0.005 g/L; Water 629 - 635 nm
Extinction Coefficient ≥14000
Wavelength 311 - 317 nm
Carbon 37.2 - 62.0 %
Nitrogen 4.8 - 8.0 %
ICP Major Analysis, Confirms Zinc Component Confirmed
Purity (HPLC) ≥75.0 %
Crystal Violet ≤0.5 %
Solubility (Colour), c = 1 mg/ml; Water Dark Blue to Very Dark Blue and Dark Blue-Green to Very Dark Blue-Green


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR