Organic reference standard, (+/-)-3-Chloro-1,2-propane-1,1,2,3,3,-d5-diol, analytical standard, Supelco®
Lieferant: Merck
32401-25MG
32401-25MGEA 343 EUR
32401-25MG
Organic reference standard, (+/-)-3-Chloro-1,2-propane-1,1,2,3,3,-d5-diol, analytical standard, Supelco®
Standards Organische Standards
3-MCPD-d5 is the deuterated form of 3-MCDP.
  • Used as an internal standard in GC-MS method for the determination of total 3-MCPD concentration from edible fats and oils.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=231b165f8b25873298adaa18a190cdf6bc3dfea2&vid=231b165f8b25873298adaa18a190cdf6bc3dfea2&prid=web&clid=SAPDAM