Organic reference standard, (+/-)-3-Chloro-1,2-propane-1,1,2,3,3,-d5-diol, analytical standard, Supelco®
Lieferant: Merck
32401-25MG
32401-25MGEA
343
EUR
32401-25MG
Organic reference standard, (+/-)-3-Chloro-1,2-propane-1,1,2,3,3,-d5-diol, analytical standard, Supelco®
Standards
Organische Standards
3-MCPD-d5 is the deuterated form of 3-MCDP.
- Used as an internal standard in GC-MS method for the determination of total 3-MCPD concentration from edible fats and oils.