2-Methyl-1-Propanol 99+% für die Spektrophotometrie
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99+%
CAS No.: 78-83-1
Molecular Formula: C4H10O
Formula Weight: 74.12
Flash Point: 28°(82°F)
Physical Form: Liquid
Density: 0.802
Refractive Index: 1.3960
Appearance: Colourless
Odour: Sweet
pH: 7
Melting Point: -108°
Boiling Point: 107-108°
Beilstein Reference: 1730878
Merck Reference: 14,5131
UN No.: 1212
MDL No.: MFCD00004740
Formel:
(CH₃)₂CHCH₂OH Siedepunkt: 107…108 °C Schmelzpunkt: –108 °C Dichte: 0,802 Flammpunkt: 28 °C (82 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004740 CAS-Nummer: 78-83-1 EINECS: 201-148-0 UN: 1212 ADR: 3,III Merck Index: 14,5131 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...