Dodecyl-β-D-glucopyranosid, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: Dodecyl-β-D-glucosid

324351-1
324351-1EA 275 EUR
324351-1
Dodecyl-β-D-glucopyranosid, Millipore®
Dodecyl-β-D-glucopyranosid
Formel: C₁₈H₃₆O₆
Molecular Weight: 348,48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00063298
CAS-Nummer: 59122-55-3
EINECS: 261-614-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR