Tetradecyltrimethylammoniumbromid ≥99% (durch Titration), Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem
Gefahr

Synonyme: Myristyltrimethylammoniumbromid, Tetradoniumbromid, TTAB, Dynamin Inhibitor II

M-Clarity™ Qualitätsstufe = MQ100

324411-500
324411-500EA 71.5 EUR
324411-500
Tetradecyltrimethylammoniumbromid ≥99% (durch Titration), Calbiochem®, Sigma-Aldrich®
Tetradecyltrimethylammoniumbromid

The Dynamin Inhibitor II, MiTMAB, also referenced under CAS 1119-97-7, controls the biological activity of Dynamin. This small molecule/inhibitor is primarily used for Membrane applications.


A cell-permeable cationic surfactant that reversibly inhibits dynamin GTPase activity (IC50 = 3.15 µM) by targeting the PH domain and interfering with phospholipid binding. Shown to block dynamin I-dependent SVE (synaptic vesicle endocytosis) in rat brain synaptosomes and dynamin II-dependent RME (receptor-mediated endocytosis) in non-neuronal HeLa, A431, and COS-7 cells. Inhibition is competitive with respect to PS and noncompetitive with respect to GTP.

Formel: C₁₇H₃₈BrN
Molecular Weight: 336,394 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00011770
CAS-Nummer: 1119-97-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR