Chloroform ≥99,5% für die Spektrophotometrie
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.5+%
CAS No.: 67-66-3
Molecular Formula: CHCl3
Formula Weight: 119.38
Physical Form: Liquid
Density: 1.492
Refractive Index: 1.4460
Odour: Characteristic
Melting Point: -63°
Boiling Point: 60.5-61.5°
Beilstein Reference: 1731042
Merck Reference: 14,2141
UN No.: 1888
MDL No.: MFCD00000826
Formel:
CHCl₃ Siedepunkt: 60,5…61.5 °C Schmelzpunkt: –63 °C Dichte: 1,492 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000826 CAS-Nummer: 67-66-3 EINECS: 200-663-8 UN: 1888 ADR: 6.1,III REACH: 05-2114614906-45 Merck Index: 14,2141 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...