Ebselen, Millipore®

Lieferant: Merck Millipore Calbiochem
Gefahr

Synonyme: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one, 2-Phenylbenzo[d][1,2]selenazol-3(2H)-one

324483-5
324483-5EA 145 EUR
324483-5
Ebselen, Millipore®
Ebselen
Formel: C₁₃H₉NOSe
Molecular Weight: 274.20 g/mol
MDL: MFCD00210937
CAS-Nummer: 60940-34-3
UN: 3283
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR