EDTA solution, 0,5 M, molecular biology grade, Calbiochem®, Millipore®
Lieferant: Merck Millipore Calbiochem
324506-100
324506-100EA
60.9
EUR
324506-100
EDTA solution, 0,5 M, molecular biology grade, Calbiochem®, Millipore®
Puffer
Klonpuffer
Sterile-filtered solution of 0,5 M EDTA in H₂O treated with diethyl pyrocarbonate (DEPC). Used in molecular biology applications.
- pH: 8,0
- Diethyl pyrocarbonate (DEPC) treated
Synonyms: Ethylenediaminetetraacetic acid trisodium salt.