Kaliumdihydrogenphosphat 98,0-100,5% (Trockenmasse) NF, FCC, J.T.Baker®

Lieferant: Avantor
Achtung

3247-05EA 311.92 EUR
3247-05 3247-07 3247-60 3247-65 3247-W3
Kaliumdihydrogenphosphat 98,0-100,5% (Trockenmasse) NF, FCC, J.T.Baker®
Kaliumdihydrogenphosphat

  • Food GMP manufactured product


No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Standards: Meets FCC and NF requirements.


Bitte beachten Store at 20° to 25°C (68° to 77°F), excursion permitted to 15° to 30°C (59° to 86°F) [see USP controlled Room Temperature].

Formel: KH₂PO₄
Siedepunkt: ∼100 °C (1013 hPa)
Schmelzpunkt: 253 °C
Dichte: 2,3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011401
CAS-Nummer: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 98.0 - 100.5%
Meets N.F. Requirements ≤3 ppm
Food GMP Manufactured Product ≤10 mg/kg
Meets F.C.C. Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical ≤0.2%
Bulk Food Chemical ≤5 ppm
NF - Assay (KH₂PO₄) (dried basis) 98.0 - 100.5 %
NF - Arsenic (As) ≤3 ppm
NF - Fluoride (F) ≤10 mg/kg
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Insoluble Substances ≤0.2 %
NF - Lead (Pb) ≤5 ppm
NF - Loss on Drying at 105°C ≤1.0 %
FCC - Assay (KH₂PO₄) (dried basis) 98.0 - 100.5 %
FCC - Arsenic (As) ≤3 ppm
FCC - Fluoride (F) ≤10 mg/kg
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Insoluble Substances ≤0.2 %
FCC - Lead (Pb) ≤2 mg/kg
FCC - Loss on Drying at 105°C ≤1.0 %
Color (APHA), For Information Only
Aluminum (Al), For Information Only
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
This product has been filled to +/- 1% of the stated net weight.
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR