Kaliumdihydrogenphosphat 98,0-100,5% (Trockenmasse) NF, FCC, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
- Food GMP manufactured product
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Standards: Meets FCC and NF requirements.
Bitte beachten Store at 20° to 25°C (68° to 77°F), excursion permitted to 15° to 30°C (59° to 86°F) [see USP controlled Room Temperature].
Formel:
KH₂PO₄ Siedepunkt: ∼100 °C (1013 hPa) Schmelzpunkt: 253 °C Dichte: 2,3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011401 CAS-Nummer: 7778-77-0 EINECS: 231-913-4 REACH: 01-2119490224-41 Merck Index: 13,07744 |
Spezifikation Testergebnisse
GMP Manufactured Product | 98.0 - 100.5% |
Meets N.F. Requirements | ≤3 ppm |
Food GMP Manufactured Product | ≤10 mg/kg |
Meets F.C.C. Requirements | Passes Test |
CAUTION: For Manufacturing, processing or repackaging | Passes Test |
Bulk Pharmaceutical Chemical | ≤0.2% |
Bulk Food Chemical | ≤5 ppm |
NF - Assay (KH₂PO₄) (dried basis) | 98.0 - 100.5 % |
NF - Arsenic (As) | ≤3 ppm |
NF - Fluoride (F) | ≤10 mg/kg |
NF - Identification A | Passes Test |
NF - Identification B | Passes Test |
NF - Insoluble Substances | ≤0.2 % |
NF - Lead (Pb) | ≤5 ppm |
NF - Loss on Drying at 105°C | ≤1.0 % |
FCC - Assay (KH₂PO₄) (dried basis) | 98.0 - 100.5 % |
FCC - Arsenic (As) | ≤3 ppm |
FCC - Fluoride (F) | ≤10 mg/kg |
FCC - Identification A | Passes Test |
FCC - Identification B | Passes Test |
FCC - Insoluble Substances | ≤0.2 % |
FCC - Lead (Pb) | ≤2 mg/kg |
FCC - Loss on Drying at 105°C | ≤1.0 % |
Color (APHA), For Information Only | |
Aluminum (Al), For Information Only | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
This product has been filled to +/- 1% of the stated net weight. | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...