Epoxomicin 95 HPLC_ASSAY_METHOD, Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem


MQ100

324800-100
324800-100EA 439 EUR
324800-100
Epoxomicin 95 HPLC_ASSAY_METHOD, Calbiochem®, Sigma-Aldrich®
Epoxomicin

An antitumor agent of microbial origin that acts as a potent, highly specific, and irreversible inhibitor of chymotrypsin-like (CT-L), trypsin-like (T-L), and peptidylglutamyl peptide hydrolyzing (PGPH) activities of the proteasome


Modifies the proteasomal catalytic subunits LMP-7, MECL1, and Z. Epoxomicin also displays anti-inflammatory activity in vivo.


Verpackung: Packaged under inert gas

Formel: C₂₈H₅₀N₄O₇
Molecular Weight: 554.7 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD03791061
CAS-Nummer: 134381-21-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR