Voriconazole, analytical standard, VETRANAL™, Supelco®
Lieferant: Merck
32483-25MG
32483-25MGEA 91.6 EUR
32483-25MG
Voriconazole, analytical standard, VETRANAL™, Supelco®
Standards Organische Standards
Voriconazole is a second-generation triazole, and is derived from fluconazole. It has a broad and enhanced antifungal spectrum as compared to other older triazoles.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8a9f4e92b4a00ea2ec5cd72309d1c71e03c823d&vid=e8a9f4e92b4a00ea2ec5cd72309d1c71e03c823d&prid=web&clid=SAPDAM