Voriconazole, analytical standard, VETRANAL™, Supelco®
Lieferant: Merck
32483-25MG
32483-25MGEA
91.6
EUR
32483-25MG
Voriconazole, analytical standard, VETRANAL™, Supelco®
Standards
Organische Standards
Voriconazole is a second-generation triazole, and is derived from fluconazole. It has a broad and enhanced antifungal spectrum as compared to other older triazoles.