Supelco® PESTANAL® Organic Reference Standard, Spirotetramat Metabolite BYI08330-Cis-Keto-Hydroxy
Lieferant: Merck
32485-10MG
32485-10MGEA
361
EUR
32485-10MG
Supelco® PESTANAL® Organic Reference Standard, Spirotetramat Metabolite BYI08330-Cis-Keto-Hydroxy
Standards
Organische Standards
- GHS signal: GHS09
Spirotetramat may be used as a reference standard for QuEChERS based methodology for the determination of spirotetramat and its relevant metabolites in pistachio samples by liquid chromatography-tandem mass spectrometry.
Bitte beachten P202 - P273 - P280 - P302 + P352 - P305 + P351 + P338 - P308 + P313