di-Kaliumhydrogenphosphat 98,0-100,5% (Trockenmasse) USP, Multikompendial endotoxinfrei, J.T.Baker®
Lieferant: Avantor
Veränderungsmanagement-Kategorie= HR
3250-01EA
190.9
EUR
3250-01
3250-05
3250-06
3250-07
3250-65
3250-60
di-Kaliumhydrogenphosphat 98,0-100,5% (Trockenmasse) USP, Multikompendial endotoxinfrei, J.T.Baker®
di-Kaliumhydrogenphosphat
- GMP manufactured product
Formel:
K₂HPO₄ Schmelzpunkt: 340 °C Dichte: 2,3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011383 CAS-Nummer: 7758-11-4 EINECS: 231-834-5 Merck Index: 13,07743 |
|
Spezifikation Testergebnisse
GMP Manufactured Product | 98.0 - 100.5% |
Meets U.S.P Requirements | ≤3 ppm |
Meets B.P. Chemical Specifications | Passes Test |
Meets E.P. Chemical Specifications | ≤0.03% |
CAUTION: For Manufacturing, processing or repackaging | Passes Test |
Bulk Pharmaceutical Chemical | ≤0.001% |
USP - Assay (K₂HPO₄) (dried basis) | 98.0 - 100.5 % |
USP - Arsenic (As) | ≤ 3 ppm |
USP - Carbonate (CO₃) | Passes Test |
USP - Chloride (Cl) | ≤ 0.03 % |
USP - Endotoxin Concentration (2.5 EU/g max.) | Passes Test |
USP - Fluoride (F) | ≤ 0.001 % |
USP - Identification A (USP) | Passes Test |
USP - Identification B (USP) | Passes Test |
USP - Insoluble Substances | ≤ 0.2 % |
USP - Iron (Fe) | ≤ 0.003 % |
USP - Loss on Drying | ≤ 1.0 % |
USP - Mono- or Tri- Basic Salt | Passes Test |
USP - pH (1 in 20) | 8.5 - 9.6 |
USP - Sodium (Na) | Passes Test |
USP - Sulfate (SO₄) | ≤ 0.1 % |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Assay (K₂HPO₄) (dried basis) | 98.0 - 101.0 % |
EP/BP - Chloride (Cl) | ≤ 200 ppm |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Identification C | Passes Test |
EP/BP - Iron (Fe) | ≤ 10 ppm |
EP/BP - Loss on Drying | ≤ 2.0 % |
EP/BP - Monopotassium phosphate | ≤ 0.025 |
EP/BP - Reducing Substances | Passes Test |
EP/BP - Sodium (Na) | ≤ 0.1 % |
EP/BP - Sulfate (SO₄) | ≤ 0.1 % |
Color (APHA), For Information Only | |
Aluminum (Al), For Information Only | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Preserve in well closed container. Store at ambient temperature. | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...