di-Kaliumhydrogenphosphat 98,0-100,5% (Trockenmasse) USP, Multikompendial endotoxinfrei, J.T.Baker®

Lieferant: Avantor


Veränderungsmanagement-Kategorie= HR

3250-01EA 190.9 EUR
3250-01 3250-05 3250-06 3250-07 3250-65 3250-60
di-Kaliumhydrogenphosphat 98,0-100,5% (Trockenmasse) USP, Multikompendial endotoxinfrei, J.T.Baker®
di-Kaliumhydrogenphosphat

  • GMP manufactured product

Formel: K₂HPO₄
Schmelzpunkt: 340 °C
Dichte: 2,3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011383
CAS-Nummer: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 98.0 - 100.5%
Meets U.S.P Requirements ≤3 ppm
Meets B.P. Chemical Specifications Passes Test
Meets E.P. Chemical Specifications ≤0.03%
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical ≤0.001%
USP - Assay (K₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Arsenic (As) ≤ 3 ppm
USP - Carbonate (CO₃) Passes Test
USP - Chloride (Cl) ≤ 0.03 %
USP - Endotoxin Concentration (2.5 EU/g max.) Passes Test
USP - Fluoride (F) ≤ 0.001 %
USP - Identification A (USP) Passes Test
USP - Identification B (USP) Passes Test
USP - Insoluble Substances ≤ 0.2 %
USP - Iron (Fe) ≤ 0.003 %
USP - Loss on Drying ≤ 1.0 %
USP - Mono- or Tri- Basic Salt Passes Test
USP - pH (1 in 20) 8.5 - 9.6
USP - Sodium (Na) Passes Test
USP - Sulfate (SO₄) ≤ 0.1 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Assay (K₂HPO₄) (dried basis) 98.0 - 101.0 %
EP/BP - Chloride (Cl) ≤ 200 ppm
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Loss on Drying ≤ 2.0 %
EP/BP - Monopotassium phosphate ≤ 0.025
EP/BP - Reducing Substances Passes Test
EP/BP - Sodium (Na) ≤ 0.1 %
EP/BP - Sulfate (SO₄) ≤ 0.1 %
Color (APHA), For Information Only
Aluminum (Al), For Information Only
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Preserve in well closed container. Store at ambient temperature.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR