di-Kaliumhydrogenphosphat 98,0-100,5% (Trockenmasse) USP, J.T.Baker®

Lieferant: Avantor

3251-07EA 1419.89 EUR
3251-07 3251-60 3251-65 3251-W3
di-Kaliumhydrogenphosphat 98,0-100,5% (Trockenmasse) USP, J.T.Baker®
di-Kaliumhydrogenphosphat

  • GMP manufactured product


No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Standards: Meets USP requirements.

Formel: K₂HPO₄
Schmelzpunkt: 340 °C
Dichte: 2,3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011383
CAS-Nummer: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product 98.0 - 100.5%
Meets U.S.P Requirements ≤3 ppm
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical ≤0.03%
USP - Assay (K₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Arsenic (As) ≤3 ppm
USP - Carbonate (CO₃) Passes Test
USP - Chloride (Cl) ≤0.03 %
USP - Fluoride (F) ≤0.001 %
USP - Identification Passes Test
USP - Insoluble Substances ≤0.2 %
USP - Iron (Fe) ≤0.003 %
USP - Loss on Drying ≤1.0 %
USP - Mono- or Tri- Basic Salt Passes Test
USP - pH (1 in 20) 8.5 - 9.6
USP - Sodium (Na) Passes Test
USP - Sulfate (SO₄) ≤0.1 %
Color (APHA), For Information Only
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR