di-Kaliumhydrogenphosphat ≥98,0% (Trockenmasse) FCC, J.T.Baker®

Lieferant: Avantor

3254-01EA 155 EUR
3254-01 3254-05
di-Kaliumhydrogenphosphat ≥98,0% (Trockenmasse) FCC, J.T.Baker®
di-Kaliumhydrogenphosphat

  • Food GMP manufactured product

Formel: K₂HPO₄
Schmelzpunkt: 340 °C
Dichte: 2,3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011383
CAS-Nummer: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets F.C.C. Requirements Passes Test
Food GMP Manufactured Product ≥98.0%
CAUTION: For Manufacturing, processing or repackaging ≤0.2%
Bulk Food Chemical ≤2.0%
FCC - Identification Passes Test
FCC - Assay (K₂HPO₄) (dried basis) ≥98.0 %
FCC - Insoluble Substances ≤0.2 %
FCC - Loss on Drying ≤2.0 %
FCC - Arsenic (As) ≤3 ppm
FCC - Fluoride (F) ≤10 ppm
FCC - Lead (Pb) ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR