Kaliumtetrahydroborat 98

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Kaliumboranat, Kaliumborhydrid, Kaliumtetrahydridoborat

32570.09 32570.22 32570.36 32570.A1
32570.09EA 18.5 EUR
32570.09 32570.22 32570.36 32570.A1
Kaliumtetrahydroborat 98
Kaliumtetrahydroborat

Purity: 98% CAS No.: 13762-51-1 Molecular Formula: BH4K Formula Weight: 53.94 Physical Form: Powder Density: 1.17 Refractive Index: 1.494 Sensitivity: Moisture Sensitive Appearance: White Odour: Odourless Melting Point: 400° dec. Solubility: Soluble in water (19g/100g ). methanol (3.9g/100g ). 4:1 water:methanol mixture (13g/100g ). Merck Reference: 14,7616 UN No.: 1870 MDL No.: MFCD00011396


Source of hydrogen, reducing agent for aldehydes, ketones, acid chlorides; foaming agent for plastics

Formel: H₄BK
Molecular Weight: 53.94 g/mol
Schmelzpunkt: >400 °C (Zers.)
Dichte: 1.17
Lagertemperatur: Raumtemperatur
MDL: MFCD00011396
CAS-Nummer: 13762-51-1
EINECS: 237-360-5
UN: 1870
ADR: 4.3 I
Merck Index: 12,07777

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR