Kresolrot ≥83% Indikator, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: o-Cresolsulphonphthalein, 3,3-Bis(4-hydroxy-3-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

32653-25G
32653-25GEA 160.11 EUR
32653-25G
Kresolrot ≥83% Indikator, Fluka™
Kresolrot
Formel: C₂₁H₁₈O₅S
Molecular Weight: 382,43 g/mol
Dichte: 0,998 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00005878
CAS-Nummer: 1733-12-6
EINECS: 217-064-2

Bestellen

Spezifikation Testergebnisse

Identity (IR) complying
Assay (HPLC) Min. 83 %
Loss on drying Max. 5 %
Change range (pH 0.5-2.5) complying
Change range (pH 6.5-8.5) complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR