Methylrot Reag. Ph. Eur. Säure-Base-Indikator, Fluka™
Lieferant: Honeywell Chemicals
Synonyme:
2-(4-(Dimethylamino)phenylazo)benzoesäure
32654-25G
32654-100G
32654-25GEA
49.33
EUR
32654-25G
32654-100G
Methylrot Reag. Ph. Eur. Säure-Base-Indikator, Fluka™
Methylrot
Formel:
C₁₅H₁₅N₃O₂ Schmelzpunkt: 179-182 °C Dichte: 0,989 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002425 CAS-Nummer: 493-52-7 EINECS: 207-776-1 Merck Index: 12,06199 |
Spezifikation Testergebnisse
Indicator solution | complying |
Test of sensitivity | complying |
Change range (pH 4.4-6.0) | complying |
UV spectrum | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...