Catecholviolett Indicator_Metal_titration, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: Pyrocatechol Violet

32672-1G 32672-5G
32672-1GEA 197.4 EUR
32672-1G 32672-5G
Catecholviolett Indicator_Metal_titration, Fluka™
Catecholviolett

Beilstein Number: 353938

Formel: C₁₉H₁₄O₇S
Molecular Weight: 386.38 g/mol
Schmelzpunkt: 185 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005868
CAS-Nummer: 115-41-3
EINECS: 204-088-3

Bestellen

Spezifikation Testergebnisse

Change range complying
Indicator Solution 0,1 % H₂O complying
Suitable for metal definition complying
Absorption maximum 436 - 446 nm
Absorbtion (1%/1cm) 400 - 500


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR