Bromphenolblau Reag. Ph. Eur. Indikator, Fluka™

Lieferant: Honeywell Chemicals
Achtung

Synonyme: 3,3',5,5'-Tetrabromphenolsulfonphthalein, 4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid

32712-5G
32712-5GEA 53.41 EUR
32712-5G
Bromphenolblau Reag. Ph. Eur. Indikator, Fluka™
Bromphenolblau
Formel: C₁₉H₁₀Br₄O₅S
Molecular Weight: 669,97 g/mol
Schmelzpunkt: 204 °C
Dichte: 1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005875
CAS-Nummer: 115-39-9
EINECS: 204-086-2
Merck Index: 13,01429

Bestellen

Spezifikation Testergebnisse

Indicator solution complying
Test of sensitivity complying
Change range (pH 2,8-4,4) complying
UV spectrum complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR