3-Amino-2-methyl-5-nitrobenzamid, Supelco®

Lieferant: Merck

32731-10MG
32731-10MGEA 367 EUR
32731-10MG
3-Amino-2-methyl-5-nitrobenzamid, Supelco®
3-Amino-2-methyl-5-nitrobenzamid
Formel: C₈H₉N₃O₃
MDL: MFCD01679715
CAS-Nummer: 3572-44-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de5fd795ec2928db97b596e8873d445c0b8a5265&vid=c7cfd161c74112595fa1b26ad042edefb953a047&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR