Eriochromschwarz T Reag. Ph. Eur. Indikator für die Metalltitration, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: 1-(2-Hydroxy-1-naphthylazo)-6-nitro-2-naphthol-4-sulfonsäure Natriumsalz, Natrium-3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalinsulfonat

32751-100G
32751-100GEA 72.36 EUR
32751-100G
Eriochromschwarz T Reag. Ph. Eur. Indikator für die Metalltitration, Fluka™
Eriochromschwarz T
Formel: C₂₀H₁₂N₃NaO₇S
Molecular Weight: 461.39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003935
CAS-Nummer: 1787-61-7
EINECS: 217-250-3
Merck Index: 13,03702

Bestellen

Spezifikation Testergebnisse

Identity (IR) complying
Solubility in methanol complying
Indicator solution complying
Suitability for Complexometry complying
UV spectrum complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR