Eriochromcyanin R S. No. 840, Fluka™

Lieferant: Honeywell Chemicals
Achtung

32752-25G
32752-25GEA 69.25 EUR
32752-25G
Eriochromcyanin R S. No. 840, Fluka™
Formel: C₂₃H₁₅Na₃O₉S
Lagertemperatur: Raumtemperatur
MDL: MFCD00012411
CAS-Nummer: 3564-18-9
EINECS: 222-641-7

Bestellen

Spezifikation Testergebnisse

Identity (IR) complying
Suitability complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR