Organic reference standard, Aflatoxin B1-13C17 solution, analytical standard, Supelco®
Lieferant: Merck
Aflatoxin B₁-¹³C₁₇ is the ¹³C isotope-labeled analog of the mycotoxin, aflatoxin.
It may be used as an analytical standard for the determination of aflatoxin B₁ in food grade gums, Pu-erh tea samples, animal feed as well as milk, milk-based infant formula, and feed samples by stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).