Bromphenolblau Natriumsalz Indikator, Fluka™
Lieferant: Honeywell Chemicals
Synonyme:
4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid Natriumsalz, Bromphenolblau wasserlöslich, 3,3',5,5'-Tetrabromphenolsulfonphthalein Natriumsalz, Natrium-α-(3,5-dibrom-4-oxo-2,5-cyclohexadienyliden)-α-(3,5-dibrom-4-hydroxyphenyl)toluolsulfonat
32768-50G
32768-50GEA
521.07
EUR
32768-50G
Bromphenolblau Natriumsalz Indikator, Fluka™
Bromphenolblau Natriumsalz
Formel:
C₁₉H₉Br₄NaO₅S Molecular Weight: 691,95 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013793 CAS-Nummer: 34725-61-6 EINECS: 252-170-2 |
Spezifikation Testergebnisse
Appearance | complying |
Identity (IR) | complying |
Clearness of the solution | complying |
Change range (pH 3-4,6) | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...