Organic reference standard, Aflatoxin B2-13C17 solution, analytical standard, Supelco®
Lieferant: Merck
32771-1ML
32771-1MLEA 2600 EUR
32771-1ML
Organic reference standard, Aflatoxin B2-13C17 solution, analytical standard, Supelco®
Standards Organische Standards
Aflatoxin B₂-¹³C₁₇ is the ¹³C isotope-labeled analog of the mycotoxin, aflatoxin.
It may be used as an analytical standard for the determination of aflatoxin B₂ in food grade gums as well as milk, milk-based infant formula, and feed samples by stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=841dac56bfdd6a6d3e8e750f043eb254a5c0629b&vid=841dac56bfdd6a6d3e8e750f043eb254a5c0629b&prid=web&clid=SAPDAM