Organic reference standard, Aflatoxin G1-13C17 solution, analytical standard, Supelco®
Lieferant: Merck
32772-1ML
32772-1MLEA
2600
EUR
32772-1ML
Organic reference standard, Aflatoxin G1-13C17 solution, analytical standard, Supelco®
Standards
Organische Standards
Aflatoxin G1-13C17 is the 13C isotope-labeled analog of the mycotoxin, aflatoxin.
It may be used as an analytical standard for the determination of aflatoxin G1 in food grade gums as well as milk, milk-based infant formula, and feed samples by stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).