Organic reference standard, Aflatoxin G₂-¹³C₁₇ solution, analytical standard, Supelco®
Lieferant: Merck
32777-1ML
32777-1MLEA 2600 EUR
32777-1ML
Organic reference standard, Aflatoxin G₂-¹³C₁₇ solution, analytical standard, Supelco®
Standards Organische Standards
Aflatoxin G₂-¹³C₁₇ is the ¹³C isotope-labeled analog of the mycotoxin, aflatoxin.
  • Analytical standard for the determination of aflatoxin G₂ in food grade gums as well as milk, milk-based infant formula
Feed samples are stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fce3fc348a235c311254e54d6fe5ec3d2452ed21&vid=fce3fc348a235c311254e54d6fe5ec3d2452ed21&prid=web&clid=SAPDAM