Organic reference standard, Aflatoxin G₂-¹³C₁₇ solution, analytical standard, Supelco®
Lieferant: Merck
32777-1ML
32777-1MLEA
2600
EUR
32777-1ML
Organic reference standard, Aflatoxin G₂-¹³C₁₇ solution, analytical standard, Supelco®
Standards
Organische Standards
Aflatoxin G₂-¹³C₁₇ is the ¹³C isotope-labeled analog of the mycotoxin, aflatoxin.
- Analytical standard for the determination of aflatoxin G₂ in food grade gums as well as milk, milk-based infant formula
Feed samples are stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).