Ethylmethylketon 99+% electronic_MOS residue free
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Ethylmethylketon, MEK, EK 500 Additiv, Methyl-ethylketon, 2-Butanon, Methylethylketon, Ethyl-methylketon
327910010
327910025
327910010.EA
49.5
EUR
327910010.
327910025.
Ethylmethylketon 99+% electronic_MOS residue free
Ethylmethylketon
Formel:
C₂H₅COCH₃ Molecular Weight: 72.11 g/mol Siedepunkt: 80 °C (760 mmHg) Schmelzpunkt: –87 °C Dichte: 0.806 g/cm³ Flammpunkt: –7 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011648 CAS-Nummer: 78-93-3 EINECS: 201-159-0 UN: 1193 ADR: 3 II REACH: 01-2119457290-43 Merck Index: 13,06097 |
|
Spezifikation Testergebnisse
Appearance | Clear liquid |
Water | ≤0.2 % (Coulometric) |
GC | ≥99.0 % |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0005 % |
Chloride (Cl) | ≤2 ppm |
Arsenic (As) | ≤20 ppb |
Barium (Ba) | ≤20 ppb |
Copper (Cu) | ≤10 ppb |
Iron (Fe) | ≤50 ppb |
Heavy metals (as Pb) | ≤1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...