Wright's Färbung solution_SC, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue

32857-1L
32857-1LEA 80.1 EUR
32857-1L
Wright's Färbung solution_SC, Sigma-Aldrich®
Wright's Färbung

Wright stain solution was used during the exudate analysis of white blood cell concentration.

Dichte: 0.8 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00082143
CAS-Nummer: 68988-92-1
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR